Learn More
4-(2-Aminoethyl)tetrahydropyran, 97%, Thermo Scientific™
Catalog Number p-7077584
Click to view available options
Quantity:
250 mg
1 g
65412-03-5
C7H16NO
130.21
MFCD02179433
BZMADPOGYCRPAI-UHFFFAOYSA-O
4-2-aminoethyl tetrahydropyran, 4-2-aminoethyl tetrahydro-2h-pyran, 2-oxan-4-yl ethan-1-amine, 2-oxan-4-yl ethanamine, 2-tetrahydro-2h-pyran-4-yl ethanamine, 4-aminoethyl-tetrahydropyrane, 2h-pyran-4-ethanamine, tetrahydro, 4-aminoethyl tetrahydropyran, 2-tetrahydro-pyran-4-yl-ethylamine, 2-tetrahydro-pyran-4-yl ethylamine
2773198
[NH3+]CCC1CCOCC1
65412-03-5
C7H16NO
130.21
MFCD02179433
BZMADPOGYCRPAI-UHFFFAOYSA-O
4-2-aminoethyl tetrahydropyran, 4-2-aminoethyl tetrahydro-2h-pyran, 2-oxan-4-yl ethan-1-amine, 2-oxan-4-yl ethanamine, 2-tetrahydro-2h-pyran-4-yl ethanamine, 4-aminoethyl-tetrahydropyrane, 2h-pyran-4-ethanamine, tetrahydro, 4-aminoethyl tetrahydropyran, 2-tetrahydro-pyran-4-yl-ethylamine, 2-tetrahydro-pyran-4-yl ethylamine
2773198
[NH3+]CCC1CCOCC1
Boiling Point 88.0°C to 89.0°C (13.0 mmHg)
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 250 mg
IUPAC Name 2-(oxan-4-yl)ethan-1-aminium
Formula Weight 129.2
Percent Purity 97%
Chemical Name or Material 4-(2-Aminoethyl)tetrahydropyran
compliance-icons
Product Identifier
  • 4-(2-Aminoethyl)tetrahydropyran, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.