missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,3,4-Tetrahydro-2-isoquinolinylmethyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7034255
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1315281-49-2
C22H28BNO2
349.281
MFCD16294539
QPDBXVVQMPYSCO-UHFFFAOYSA-N
2-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-1,2,3,4-tetrahydro-isoquinoline, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-1,2,3,4-tetrahydroisoquinoline, 4-1,2,3,4-tetrahydro-2-isoquinolinylmethyl benzeneboronic acid pinacol ester, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl-3,4-dihydro-1h-isoquinoline, amtb238, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl-1,2,3,4-tetrahydroisoquinoline
71306530
2-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]-3,4-dihydro-1H-isoquinoline
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCC4=CC=CC=C4C3
This item is not returnable. View return policy
1315281-49-2
C22H28BNO2
349.281
MFCD16294539
QPDBXVVQMPYSCO-UHFFFAOYSA-N
2-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-1,2,3,4-tetrahydro-isoquinoline, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-1,2,3,4-tetrahydroisoquinoline, 4-1,2,3,4-tetrahydro-2-isoquinolinylmethyl benzeneboronic acid pinacol ester, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl-3,4-dihydro-1h-isoquinoline, amtb238, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl-1,2,3,4-tetrahydroisoquinoline
71306530
2-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]-3,4-dihydro-1H-isoquinoline
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCC4=CC=CC=C4C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 349.28
Percent Purity 95%
Chemical Name or Material 4-(1,2,3,4-Tetrahydro-2-isoquinolinylmethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'