Learn More
(3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile Hydrochloride (Isodidiavalo Hydrochloride), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
(3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile Hydrochloride (Isodidiavalo Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | (3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile Hydrochloride (Isodidiavalo Hydrochloride) |
| CAS | 63867-26-5 |
| Analyte or Component Names | (3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile Hydrochloride (Isodidiavalo Hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 N2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methadone Hydrochoride Imp. C (EP), Methadone Imp. C (EP), (3RS)-4-(Dimethylamino)-3-methyl-2,2-diphenylbutanenitrile Hydrochloride, Methadone Hydrochoride Imp. C (EP) as Hydrochloride, Isodidiavalo Hydrochloride, Methadone Related Compound C as Hydrochloride, Methadone Hydrochloride Impurity C as Hydrochloride, Methadone Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C19H22N2.ClH/c1-16(14-21(2)3)19(15-20,17-10-6-4-7-11-17)18-12-8-5-9-13-18;/h4-13,16H,14H2,1-3H3;1H |
| SMILES | Cl.CC(CN(C)C)C(C#N)(c1ccccc1)c2ccccc2 |
| IUPAC Name | 4-(dimethylamino)-3-methyl-2,2-diphenylbutanenitrile;hydrochloride |
| Show More |