Learn More
Ratiolab X250 Container PP Natural 120ml w/Red Screw Cap Sterile
Brand:  Ratiolab 5921183
X250 Container PP Natural 120ml w/Red Screw Cap Sterile
Product Code. 30968819
  • ONLINE EXCLUSIVE PRICE
    955.00 NOK
    942.35 NOK / Pack of 250
    Save 12.65 NOK
    1% Off
Estimated Shipment: 21-12-2026