Learn More
TET2 Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35520JF549
Product Code. 30500702
  • 6175.00 NOK / 0.10mL
Description
TET2 Polyclonal antibody specifically detects TET2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TET2
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
54790
Store at 4°C in the dark.
Purified
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Janelia Fluor 549
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
Recombinant fusion protein containing a sequence corresponding to amino acids 1833-2002 of human TET2 (NP_001120680.1).,, Sequence:, VQGVASGAEDNDEVWSDSEQSFLDPDIGGVAVAPTHGSILIECAKRELHATTPLKNPNRNHPTRISLVFYQHKSMNEPKHGLALWEAKMAEKAREKEEECEKYGPDYVPQKSHGKKVKREPAEPHETSEPTYLRFIKSLAERTMSVTTDSTVTTSPYAFTRVTGPYNRYI
0.1 mL
Primary
Human, Mouse, Rat