Learn More
Spike Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-37899MFV610
Product Code. 30500687
  • 6565.00 NOK / 0.10mL
Description
Spike Polyclonal antibody specifically detects Spike in SARS-CoV samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spike
Polyclonal
50mM Sodium Borate
A synthetic peptide corresponding to a sequence within amino acids 600-700 of coronavirus Spike (NP_828851.1).,, Sequence:, DVNCTDVSTAIHADQLTPAWRIYSTGNNVFQTQAGCLIGAEHVDTSYECDIPIGAGICASYHTVSLLRSTSQKSIVAYTMSLGADSSIAYSNNTIAIPTNF
0.1 mL
Primary
Store at 4°C in the dark.
Purified
ELISA, Western Blot
mFluor Violet 610 SE
Rabbit
Affinity purified
RUO
SARS-CoV