missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3CA Antibody [PE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37884PE
This item is not returnable.
View return policy
Description
PIK3CA Polyclonal antibody specifically detects PIK3CA in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PIK3CA | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| PE | |
| EC 2.7.1, EC 2.7.1.153, MGC142161, MGC142163, p110-alpha, Phosphatidylinositol 3-Kinase catalytic subunit alpha, phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha, phosphatidylinositol 3-kinase, catalytic, alpha polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform, phosphoinositide-3-kinase, catalytic, alpha polypeptide, PI3K, PI3K-alpha, PI3-kinase p110 subunit alpha, PI3-kinase subunit alpha, PtdIns-3-kinase p110, PtdIns-3-kinase subunit alpha, PtdIns-3-kinase subunit p110-alpha | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-600 of human PIK3CA (NP_006209.2).,, Sequence:, DCFTMPSYSRRISTATPYMNGETSTKSLWVINSALRIKILCATYVNVNIRDIDKIYVRTGIYHGGEPLCDNVNTQRVPCSNPRWNEWLNYDIYIPDLPRAARLCLSICSVKGRKGAKEEHCPLAWGNINLFDYTDTLVSGKMALNLWPVPHGLEDLLNPIGVTGSNPNKETPCLELEFDWFSSVVKFPDMSVIEEHANWSVSREAGFSYSHAGLSNRLARDNELRENDKEQLKAISTRDPLSEITEQEKDFLWSHRHYCVTIPEILPKLLLSVKWNSRDEVAQMYCLVKDWPPIKPEQAME | |
| 0.1 mL | |
| Breast Cancer, Cancer, Diabetes Research, mTOR Pathway, Oncogenes | |
| 5290 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction