missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD2 Antibody [DyLight 680], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35216FR
This item is not returnable.
View return policy
Description
SMARCD2 Polyclonal antibody specifically detects SMARCD2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SMARCD2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 60 kDa BRG-1/Brm-associated factor subunit B, BAF60B, BRG1-associated factor 60B, chromatin remodeling complex BAF60B subunit, CRACD2, mammalian chromatin remodeling complex BRG1-associated factor 60B, PRO2451, Rsc6p, SWI/SNF complex 60 kDa subunit B, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2, Swp73-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 454-531 of human SMARCD2 (NP_001091896.1).,, Sequence:, RDFMLSFSTDPQDFIQEWLRSQRRDLKIITDVIGNPEEERRAAFYHQPWAQEAVGRHIFAKVQQRRQELEQVLGIRLT | |
| 0.1 mL | |
| Chromatin Research | |
| 6603 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction