missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF1 Antibody [DyLight 680], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35204FR
This item is not returnable.
View return policy
Description
RASSF1 Polyclonal antibody specifically detects RASSF1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| RASSF1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| DyLight 680 | |
| 123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 101-340 of human RASSF1 (NP_009113.3).,, Sequence:, CCGPRDLGWEPAVERDTNVDEPVEWETPDLSQAEIEQKIKEYNAQINSNLFMSLNKDGSYTGFIKVQLKLVRPVSVPSSKKPPSLQDARRGPGRGTSVRRRTSFYLPKDAVKHLHVLSRTRAREVIEALLRKFLVVDDPRKFALFERAERHGQVYLRKLLDDEQPLRLRLLAGPSDKALSFVLKENDSGEVNWDAFSMPELHNFLRILQREEEEHLRQILQKYSYCRQKIQEALHACPLG | |
| 0.1 mL | |
| Breast Cancer, Cell Cycle and Replication, Mitotic Regulators | |
| 11186 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction