missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOX Antibody [Alexa Fluor« 750], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-27880AF750
This item is not returnable.
View return policy
Description
TOX Polyclonal antibody specifically detects TOX in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| TOX | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 750 | |
| KIAA0808TOX1, thymocyte selection-associated high mobility group box, thymocyte selection-associated high mobility group box protein TOX, Thymus high mobility group box protein TOX | |
| TOX antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human TOX. The immunogen is located within amino acids 130 - 180 of TOX. | |
| 0.1 mL | |
| Breast Cancer | |
| 9760 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction