Learn More
PLK4 Antibody [HRP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-37985H
Product Code. 30500596
  • 6175.00 NOK / 0.10mL
Description
PLK4 Polyclonal antibody specifically detects PLK4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLK4
Polyclonal
PBS
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
HRP
EC 2.7.11.21, PLK-4, polo-like kinase 4 (Drosophila), polo-like kinase 4SAKSTK18serine/threonine kinase 18, Sak, serine/threonine protein kinase SAK, Serine/threonine-protein kinase 18, serine/threonine-protein kinase PLK4, Serine/threonine-protein kinase Sak, Snk akin kinase
Recombinant fusion protein containing a sequence corresponding to amino acids 641-970 of human PLK4 (NP_055079.3).,, Sequence:, SDGNTITIYYPNGGRGFPLADRPPSPTDNISRYSFDNLPEKYWRKYQYASRFVQLVRSKSPKITYFTRYAKCILMENSPGADFEVWFYDGVKIHKTEDFIQVIEKTGKSYTLKSESEVNSLKEEIKMYMDHANEGHRICLALESIISEEERKTRSAPFFPIIIGRKPGSTSSPKALSPPPSVDSNYPTRERASFNRMVMHSAASPTQAPILNPSMVTNEGLGLTTTASGTDISSNSLKDCLPKSAQLLKSVFVKNVGWATQLTSGAVWVQFNDGSQLVVQAGVSSISYTSPNGQTTRYGENEKLPDYIKQKLQCLSSILLMFSNPTPNFH
0.1 mL
Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers
10733
Store at 4°C in the dark.