missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumarylacetoacetate hydrolase Antibody [Alexa Fluor« 700], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35351AF700
This item is not returnable.
View return policy
Description
Fumarylacetoacetate hydrolase Polyclonal antibody specifically detects Fumarylacetoacetate hydrolase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Fumarylacetoacetate hydrolase | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Alexa Fluor 700 | |
| beta-diketonase, EC 3.7.1.2, FAA, fumarylacetoacetase, Fumarylacetoacetate hydrolase, fumarylacetoacetate hydrolase (fumarylacetoacetase) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-419 of human Fumarylacetoacetate hydrolase (NP_000128.1).,, Sequence:, MSFIPVAEDSDFPIHNLPYGVFSTRGDPRPRIGVAIGDQILDLSIIKHLFTGPVLSKHQDVFNQPTLNSFMGLGQAAWKEARVFLQNLLSVSQARLRDDTELRKCAFISQASATMHLPATIGDYTDFYSSRQHATNVGIMFRDKENALMPNWLHLPVGYHGRASSVVVSGTPIRRPMGQMKPDDSKPPVYGACKLLDMELEMAFFVGPGNRLGEPIPISKAHEHIFGMVLMNDWSARDIQKWEYVPLGPFLGKSFGTTVSPWVVPMDALMPFAVPNPKQDPRPLPYLCHDEPYTFDINLSVNLKGEGMSQAATICKSNFKYMYWTMLQQLTHHSVNGCNLRPGDLLASGTISGPEPENFGSMLELSWKGTKPIDLGNGQTRKFLLDGDEVIITGYCQGDGYRIGFGQCAGKVLPALLPS | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 2184 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction