missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2G2 Antibody [DyLight 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35140G
This item is not returnable.
View return policy
Description
UBE2G2 Polyclonal antibody specifically detects UBE2G2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| UBE2G2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 488 | |
| EC 6.3.2.19, UBC7human ubiquitin conjugating enzyme G210ubiquitin-conjugating enzyme E2G 2 (homologous to yeast UBC7), Ubiquitin carrier protein G2, ubiquitin conjugating enzyme 7, ubiquitin conjugating enzyme G2, ubiquitin-conjugating enzyme E2 G2, ubiquitin-conjugating enzyme E2G 2 (UBC7 homolog, yeast), Ubiquitin-protein ligase G2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-165 of human UBE2G2 (NP_003334.2).,, Sequence:, MAGTALKRLMAEYKQLTLNPPEGIVAGPMNEENFFEWEALIMGPEDTCFEFGVFPAILSFPLDYPLSPPKMRFTCEMFHPNIYPDGRVCISILHAPGDDPMGYESSAERWSPVQSVEKILLSVVSMLAEPNDESGANVDASKMWRDDREQFYKIAKQIVQKSLGL | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 7327 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction