Learn More
TREM2 Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35146AF532
Product Code. 30500490
  • 6175.00 NOK / 0.10mL
Description
TREM2 Polyclonal antibody specifically detects TREM2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TREM2
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Alexa Fluor 532
Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Recombinant fusion protein containing a sequence corresponding to amino acids 19-161 of human TREM2 (NP_061838.1).,, Sequence:, HNTTVFQGVAGQSLQVSCPYDSMKHWGRRKAWCRQLGEKGPCQRVVSTHNLWLLSFLRRWNGSTAITDDTLGGTLTITLRNLQPHDAGLYQCQSLHGSEADTLRKVLVEVLADPLDHRDAGDLWFPGESESFEDAHVEHSISR
0.1 mL
Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience
54209
Store at 4°C in the dark.