missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p57 Kip2 Antibody [Alexa Fluor« 700], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-27877AF700
This item is not returnable.
View return policy
Description
p57 Kip2 Polyclonal antibody specifically detects p57 Kip2 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| p57 Kip2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 700 | |
| Beckwith-Wiedemann syndrome, BWS, cyclin-dependent kinase inhibitor 1C, cyclin-dependent kinase inhibitor 1C (p57, Kip2), Cyclin-dependent kinase inhibitor p57, KIP2BWCR, p57, p57Kip2, WBS | |
| p57 Kip2 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human p57 Kip2. The immunogen is located within amino acids 40 - 90 of p57 Kip2. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 1028 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction