missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1C2 Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35145AF594
This item is not returnable.
View return policy
Description
AKR1C2 Polyclonal antibody specifically detects AKR1C2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| AKR1C2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1646 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 594 | |
| aldo-keto reductase family 1 member C2, aldo-keto reductase family 1, member C2 (dihydrodiol dehydrogenase 2; bile acidbinding protein; 3-alpha hydroxysteroid dehydrogenase, type III), BABP, Chlordecone reductase homolog HAKRD, DD, DD-2, DD2DD/BABP, DDH2FLJ53800, Dihydrodiol dehydrogenase 2, Dihydrodiol dehydrogenase/bile acid-binding protein, EC 1.1.1,3-alpha-HSD3, EC 1.1.1.213, EC 1.3.1.20, HAKRDAKR1C-pseudo, HBAB, MCDR2, pseudo-chlordecone reductase, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II dihydrodiol dehydrogenase, Type III 3-alpha-hydroxysteroid dehydrogenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human AKR1C2 (NP_995317.1).,, Sequence:, MDSKYQCVKLNDGHFMPVLGFGTYAPAEVPKSKALEAVKLAIEAGFHHIDSAHVYNNEEQVGLAIRSKIADGSVKREDIFYTSKLWSNSHRPELVRPALERSLKNLQLDYVDLYLIHFPVSVKPGEEVIPKDENGKILFDTVDLCATWEAMEKCKDAGLAKSIGVSNFNHRLLEMILNKPGLKYKPVCNQVECHPYFNQRKLLDFCKSKDIVLVAYSALGSHREEPWVDPNSPVLLEDPVLCALAKKHKRTPALIALRYQLQRGVVVLAKSYNEQRIRQNVQVFEFQLTSEEMKAIDGLNRNVRYLTLDIFAGPPNYPFSDEY | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction