missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASK Antibody [DyLight 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38000G
This item is not returnable.
View return policy
Description
CASK Polyclonal antibody specifically detects CASK in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| CASK | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 488 | |
| Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase, CAMGUK, CMG, EC 2.7.11, EC 2.7.11.1, FGS4CAGH39, FLJ22219, hCASK, LIN2FLJ31914, MICPCH, peripheral plasma membrane protein CASK, Protein lin-2 homolog, TNRC8, trinucleotide repeat containing 8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CASK (NP_001119526.1).,, Sequence:, MADDDVLFEDVYELCEVIGKGPFSVVRRCINRETGQQFAVKIVDVAKFTSSPGLSTEDLKREASICHMLKHPHIVELLETYSSDGMLYMVFEFMDGADLCFEIVKRADAGFVYSEAVASHYMRQILEALRYCHDNNIIHRDVKPHCVLLASKENSAPVKLGGFGVAIQLGESGLVAGGRVGTPHFMAPEVVKREPYGKPVDVWGCGVILFILLSGCLPFYGTKERLFEGIIKGKYKMNPRQWSHISESAKDLVRRMLMLDPAERITVYEALNHPWLKERDRYAYKIHLPETVEQLRKFNA | |
| 0.1 mL | |
| Signal Transduction | |
| 8573 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction