Learn More
RASSF1 Antibody [Biotin], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35204B
Product Code. 30500384
  • 6175.00 NOK / 0.10mL
Description
RASSF1 Polyclonal antibody specifically detects RASSF1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RASSF1
Polyclonal
PBS
Rabbit
Affinity purified
RUO
Primary
Human
Antibody
IgG
ELISA, Western Blot
Biotin
123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5
Recombinant fusion protein containing a sequence corresponding to amino acids 101-340 of human RASSF1 (NP_009113.3).,, Sequence:, CCGPRDLGWEPAVERDTNVDEPVEWETPDLSQAEIEQKIKEYNAQINSNLFMSLNKDGSYTGFIKVQLKLVRPVSVPSSKKPPSLQDARRGPGRGTSVRRRTSFYLPKDAVKHLHVLSRTRAREVIEALLRKFLVVDDPRKFALFERAERHGQVYLRKLLDDEQPLRLRLLAGPSDKALSFVLKENDSGEVNWDAFSMPELHNFLRILQREEEEHLRQILQKYSYCRQKIQEALHACPLG
0.1 mL
Breast Cancer, Cell Cycle and Replication, Mitotic Regulators
11186
Store at 4°C in the dark.