missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP98 Antibody [Alexa Fluor« 350], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35075AF350
This item is not returnable.
View return policy
Description
NUP98 Polyclonal antibody specifically detects NUP98 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| NUP98 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 350 | |
| ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 621-920 of human NUP98 (NP_624357.1).,, Sequence:, SKPVDENHQQDGDEDSLVSHFYTNPIAKPIPQTPESAGNKHSNSNSVDDTIVALNMRAALRNGLEGSSEETSFHDESLQDDREEIENNSYHMHPAGIILTKVGYYTIPSMDDLAKITNEKGECIVSDFTIGRKGYGSIYFEGDVNLTNLNLDDIVHIRRKEVVVYLDDNQKPPVGEGLNRKAEVTLDGVWPTDKTSRCLIKSPDRLADINYEGRLEAVSRKQGAQFKEYRPETGSWVFKVSHFSKYGLQDSDEEEEEHPSKTSTKKLKTAPLPPASQTTPLQMALNGKPAPPPQVEKKGQ | |
| 0.1 mL | |
| Cancer, Mitotic Regulators | |
| 4928 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction