Learn More
PICK1 Antibody [Biotin], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35510B
Product Code. 30500353
  • 6175.00 NOK / 0.10mL
Description
PICK1 Polyclonal antibody specifically detects PICK1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PICK1
Polyclonal
PBS
Rabbit
Affinity purified
RUO
Primary
Human
Antibody
IgG
ELISA, Western Blot
Biotin
alpha binding protein, dJ1039K5, MGC15204, PICK, PRKCA-binding protein, PRKCABPprotein interacting with PRKCA, Protein interacting with C kinase 1, protein interacting with PRKCA 1, Protein kinase C-alpha-binding protein
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human PICK1 (NP_036539.1).,, Sequence:, YLSYCLKVKEMDDEEYSCIALGEPLYRVSTGNYEYRLILRCRQEARARFSQMRKDVLEKMELLDQKHVQDIVFQLQRLVSTMSKYYNDCYAVLRDADVFPI
0.1 mL
Signal Transduction, Transcription Factors and Regulators, Vision
9463
Store at 4°C in the dark.