Learn More
PDLIM4 Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-37889AF488
Product Code. 30500333
  • 6175.00 NOK / 0.10mL
Description
PDLIM4 Polyclonal antibody specifically detects PDLIM4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDLIM4
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
8572
Store at 4°C in the dark.
Purified
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Alexa Fluor 488
LIM domain protein, LIM protein RIL, PDZ and LIM domain 4, PDZ and LIM domain protein 4, Reversion-induced LIM protein, RILenigma homolog
Recombinant fusion protein containing a sequence corresponding to amino acids 80-210 of human PDLIM4 (NP_003678.2).,, Sequence:, VSRPEGRSWPSAPDDSKAQAHRIHIDPEIQDGSPTTSRRPSGTGTGPEDGRPSLGSPYGQPPRFPVPHNGSSEATLPAQMSTLHVSPPPSADPARGLPRSRDCRVDLGSEVYRMLREPAEPVAAEPKQSGS
0.1 mL
Primary
Human, Mouse, Rat