missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin 1/AQP1 Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35493JF669
This item is not returnable.
View return policy
Description
Aquaporin 1/AQP1 Polyclonal antibody specifically detects Aquaporin 1/AQP1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Aquaporin 1/AQP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 669 | |
| 28-kDa, AQP-1, AQP-CHIP, aquaporin 1 (channel-forming integral protein, 28kDa), aquaporin 1 (Colton blood group), Aquaporin-CHIP, CHIP2828kDa, CO blood group), CO, MGC26324 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-269 of human Aquaporin 1/AQP1 (AQP1) (NP_932766.1).,, Sequence:, AVITHNFSNHWIFWVGPFIGGALAVLIYDFILAPRSSDLTDRVKVWTSGQVEEYDLDADDINSRVEMKPK | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 358 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction