missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad9 [p Tyr92] Antibody (ARC65536) [CoraFluorÖ 1], Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33580CL1
This item is not returnable.
View return policy
Description
Smad9 Monoclonal antibody specifically detects Smad9 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Smad9 | |
| Monoclonal | |
| CoraFluor 1 | |
| MAD homolog 9, MAD, mothers against decapentaplegic homolog 9 (Drosophila), Madh6, MADH6mothers against decapentaplegic homolog 9, MADH9Mothers against decapentaplegic, drosophila, homolog of, 9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9SMAD8B, SMAD, mothers against DPP homolog 9, SMAD, mothers against DPP homolog 9 (Drosophila), SMAD8, SMAD8A, Smad9 | |
| A synthetic phosphorylated peptide around Y92 of human Smad9.1). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| ARC65536 | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4093 | |
| Store at 4°C in the dark. Do not freeze. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction