missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centrin 1 Antibody [DyLight 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38050G
This item is not returnable.
View return policy
Description
Centrin 1 Polyclonal antibody specifically detects Centrin 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Centrin 1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| DyLight 488 | |
| calcium binding protein, caltractin, Caltractin isoform 2, CEN1centrin-1, centrin, EF-hand protein, 1, CETN, EF-hand protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human Centrin 1 (NP_004057.1).,, Sequence:, MASGFKKPSAASTGQKRKVAPKPELTEDQKQEVREAFDLFDVDGSGTIDAKELKVAMRALGFEPRKEEMKKMISEVDREGTGKISFNDFLAVMTQKMSEKDTKEEILKAFRLFDDDETGKISFKNLKRVANELGENLTDEELQEMIDEADRDGDGEVNEEEFLRIMKKTSLY | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1068 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction