missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USF1 Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35360MFV500
This item is not returnable.
View return policy
Description
USF1 Polyclonal antibody specifically detects USF1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| USF1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| mFluor Violet 500 SE | |
| BHLHB11, bHLHb11FCHL1, Class B basic helix-loop-helix protein 11, FCHL, HYPLIP1, Major late transcription factor 1, MLTF, MLTFI, UEF, upstream stimulatory factor 1, upstream transcription factor 1, USF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human USF1 (NP_009053.1).,, Sequence:, MKGQQKTAETEEGTVQIQEGAVATGEDPTSVAIASIQSAATFPDPNVKYVFRTENGGQVMYRVIQVSEGQLDGQTEGTGAISGYPATQSMTQAVIQGAFTSDDAVDTEGTAAETHYTYFPSTAVGDGAGGTTSGSTAAVVTTQGSEALLGQATPPGTGQFFVMMSPQEVLQGGSQRSIAPRTHPYSPKSEAPRTTRDEKRRAQHNEVERRRRDKINNWIVQLSKIIPDCSMESTKSGQSKGGILSKACDYIQELRQSNHRLSEELQGLDQLQLDNDVLRQQVEDLKNKNLLLRAQLRHHGLEVVIKNDSN | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 7391 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction