missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37899JF549
This item is not returnable.
View return policy
Description
Spike Polyclonal antibody specifically detects Spike in SARS-CoV samples. It is validated for ELISA,Western Blot
Specifications
| Spike | |
| Polyclonal | |
| 50mM Sodium Borate | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of coronavirus Spike (NP_828851.1).,, Sequence:, DVNCTDVSTAIHADQLTPAWRIYSTGNNVFQTQAGCLIGAEHVDTSYECDIPIGAGICASYHTVSLLRSTSQKSIVAYTMSLGADSSIAYSNNTIAIPTNF | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| Janelia Fluor 549 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| SARS-CoV | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction