missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GO Protein alpha Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38002MFV610
This item is not returnable.
View return policy
Description
GO Protein alpha Polyclonal antibody specifically detects GO Protein alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| GO Protein alpha | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| mFluor Violet 610 SE | |
| DKFZp686O0962, G-ALPHA-o, GNAO, guanine nucleotide binding protein (G protein), alpha activating activitypolypeptide O, guanine nucleotide binding protein, alpha activating polypeptide O, guanine nucleotide-binding protein G(o) subunit alpha | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human GO Protein alpha (NP_066268.1).,, Sequence:, GFSGEDVKQYKPVVYSNTIQSLAAIVRAMDTLGIEYGDKERKADAKMVCDVVSRMEDTEPFSAELLSAMMRLWGDSGIQECFNRSREYQLNDSAKYYLDSLDRIGAADYQPTEQDILRTRV | |
| 0.1 mL | |
| Signal Transduction | |
| 2775 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction