missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGPS1 Antibody [Janelia Fluor« 525], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35656JF525
This item is not returnable.
View return policy
Beskrivelse
GGPS1 Polyclonal antibody specifically detects GGPS1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Tekniske data
| GGPS1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Janelia Fluor 525 | |
| 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, EC 2.5.1.29, Farnesyl diphosphate synthase, Farnesyltranstransferase, Geranylgeranyl diphosphate synthase, geranylgeranyl diphosphate synthase 1, geranylgeranyl pyrophosphate synthase, geranyltranstransferase, GGPP synthase, GGPPS, GGPPS1farnesyltranstransferase, GGPPSase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 164-300 of human GGPS1 (NP_001032354.1).,, Sequence:, QLFSDYKEDLKPLLNTLGLFFQIRDDYANLHSKEYSENKSFCEDLTEGKFSFPTIHAIWSRPESTQVQNILRQRTENIDIKKYCVHYLEDVGSFEYTRNTLKELEAKAYKQIDARGGNPELVALVKHLSKMFKEENE | |
| 0.1 mL | |
| Cancer | |
| 9453 | |
| Store at 4°C in the dark. | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion