missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTRF Antibody [DyLight 594], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35822DL594
This item is not returnable.
View return policy
Description
PTRF Polyclonal antibody specifically detects PTRF in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| PTRF | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| DyLight 594 | |
| CAVIN, Cavin-1, CGL4, FLJ90031, polymerase I and transcript release factor, RNA polymerase I and transcript release factor, TTF-I interacting peptide 12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-280 of human PTRF (NP_036364.2).,, Sequence:, ELSSDEAVEVEEVIEESRAERIKRSGLRRVDDFKKAFSKEKMEKTKVRTRENLEKTRLKTKENLEKTRHTLEKRMNKLGTR | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism | |
| 284119 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction