missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAF35 Antibody [Janelia Fluor« 646], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37920JF646
This item is not returnable.
View return policy
Description
BRAF35 Polyclonal antibody specifically detects BRAF35 in Human samples. It is validated for ELISA,Western Blot
Specifications
| BRAF35 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Janelia Fluor 646 | |
| BRAF35Sox-like transcriptional factor, high-mobility group 20B, HMG domain-containing protein 2, HMG domain-containing protein HMGX2, HMGX2FLJ26127, HMGXB2member 1-related, SMARCE1R, SMARCE1-related protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human BRAF35 (NP_006330.2).,, Sequence:, MSHGPKQPGAAAAPAGGKAPGQHGGFVVTVKQERGEGPRAGEKGSHEEEPVKKRGWPKGKKRKKILPNGPKAPVTGYVRFLNERREQIRTRHPDLPFPEITKMLGAEWSKLQPTEKQRYLDEAEREKQQYMKELRAYQQSEAYKMCTEKIQEKKIKKEDSSSGLMNTLLNGHKGGDCDGFSTFDVPIFTEEFLDQNKAREAELRRLRKMNVAFEEQNAVLQRHTQSMSSARERLEQELALEERRTLALQQQLQAVRQALTASFASLPVPGTGETPTLGTLDFYMARLHGAIERDPAQHEKLIVRIKEILAQVASEHL | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 10362 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction