missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPI17 alpha Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35103AF405
This item is not returnable.
View return policy
Description
CPI17 alpha Polyclonal antibody specifically detects CPI17 alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CPI17 alpha | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Alexa Fluor 405 | |
| 17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human CPI17 alpha (NP_150281.1).,, Sequence:, MAAQRLGKRVLSKLQSPSRARGPGGSPGGLQKRHARVTVKYDRRELQRRLDVEKWIDGRLEELYRGMEADMPDEINIDELLELESEEERSRKIQGLLKSCGKPVEDFIQELLAKLQGLHRQPGLRQPSPSHDGSLSPLQDRARTAHP | |
| 0.1 mL | |
| Phospho Specific, Protein Phosphatase | |
| 94274 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction