missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoleucyl tRNA synthetase Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35104JF669
This item is not returnable.
View return policy
Description
Isoleucyl tRNA synthetase Polyclonal antibody specifically detects Isoleucyl tRNA synthetase in Human samples. It is validated for ELISA,Western Blot
Specifications
| Isoleucyl tRNA synthetase | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Janelia Fluor 669 | |
| EC 6.1.1, EC 6.1.1.5, FLJ20736, IARS1, ILERS, ILRS, IRS, isoleucine tRNA ligase 1, cytoplasmic, Isoleucine--tRNA ligase, isoleucyl-tRNA synthetase, isoleucyl-tRNA synthetase, cytoplasmic, PRO0785 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1158-1262 of human Isoleucyl tRNA synthetase (NP_002152.2).,, Sequence:, SAPSLINSSSTLLCQYINLQLLNAKPQECLMGTVGTLLLENPLGQNGLTHQGLLYEAAKVFGLRSRKLKLFLNETQTQEITEDIPVKTLNMKTVYVSVLPTTADF | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 3376 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction