missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELAVL4 Antibody [Biotin], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35160B
This item is not returnable.
View return policy
Description
ELAVL4 Polyclonal antibody specifically detects ELAVL4 in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| ELAVL4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Biotin | |
| abnormal vision, Drosophila, homolog of, like-4, ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D), HuD, Paraneoplastic encephalomyelitis antigen HuD | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ELAVL4 (NP_068771.2).,, Sequence:, MVMIISTMEPQVSNGPTSNTSNGPSSNNRNCPSPMQTGATTDDSKTNLIVNYLPQNMTQEEFRSLFGSIGEIESCKLVRDKITGQSLGYGFVNYIDPKDA | |
| 0.1 mL | |
| Neuroscience, Vision | |
| 1996 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction