missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCX Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35594AF594
This item is not returnable.
View return policy
Description
SMCX Polyclonal antibody specifically detects SMCX in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SMCX | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8242 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| Alexa Fluor 594 | |
| DXS1272EMRXJ, EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1C, JARID1Clysine-specific demethylase 5C, jumonji, AT rich interactive domain 1C, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, lysine (K)-specific demethylase 5C, MRXSJ, Protein SmcX, Protein Xe169, Smcx homolog, X chromosome, SMCXselected cDNA on X, Smcy homolog, X-linked, Smcy homolog, X-linked (mouse), XE169JmjC domain-containing protein SMCX | |
| A synthetic peptide corresponding to a sequence within amino acids 1400 to the C-terminus of human SMCX (XP_011529128.1).,, Sequence:, ERHGSRARGRALERRRRRKVDRGGEGDDPAREELEPKRVRSSGPEAEEVQEEEELEEETGGEGPPAPIPTTGSPSTQENQNGLEPAEGTTSGPSAPFSTLTPRLHLPCPQQPPQQQL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction