Learn More
gamma Tubulin Antibody (6T10A1) [Janelia Fluor« 646], Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-33210JF646
Product Code. 30500002
  • 5780.00 NOK / 0.10mL
Description
gamma Tubulin Monoclonal antibody specifically detects gamma Tubulin in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
gamma Tubulin
Monoclonal
Janelia Fluor 646
gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide
Recombinant protein of human gamma Tubulin.,, Sequence:, MPREIITLQLGQCGNQIGFEFWKQLCAEHGISPEGIVEEFATEGTDRKDVFFYQADDEHYIPRAVLLDLEPRVIHSILNSPYAKLYNPENIYLSEHGGGAGNNWASGFSQGEKIHEDIFDIIDREADGSDSLEGFVLCHSIAGGTGSGLGSYLLERLNDRYPKKLVQTYSVFPNQDEMSDVVVQPYNSLLTLKRLTQNADCVVVLDNTALNRIATDRLHIQNPSFSQINQLVSTIMSASTTTLRYPGYMNNDLIGLIASLIPTPRLHFLMTGYTPLTTDQSVASVRKTTVLDVMRRLLQPKNVMVSTGRDRQTNHCYIAILNIIQGEVDPTQVHKSLQRIRERKLANFIPWGPASIQVALSRKSPYLPSAHRVSGLMMANHTSISSLFERTCRQYDKLRKREAFLEQFRKEDMFKDNFDEMDTSREIVQQLIDEYHAATRPDYISWGTQEQ
0.1 mL
Cell Biology, Cellular Markers, Centrosome Markers
7283
Store at 4°C in the dark.
Purified
ELISA, Immunoprecipitation, Western Blot
6T10A1
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat