missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycogen Phosphorylase BB/GPBB Antibody (E9B10) [Alexa Fluor« 350], Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-27945AF350
This item is not returnable.
View return policy
Description
Glycogen Phosphorylase BB/GPBB Monoclonal antibody specifically detects Glycogen Phosphorylase BB/GPBB in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Glycogen Phosphorylase BB/GPBB | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Brain glycogen phosphorylase, EC 2.4.1.1, Glycogen phosphorylase B, Glycogen phosphorylase brain form, Glycogen Phosphorylase Isoenzyme BB, MGC9213, Phosphorylase glycogen brain, PYGB | |
| Synthetic peptide derived from human Glycogen Phosphorylase BB/GPBB | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| E9B10 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5834 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction