Learn More
KLF1 Antibody [Alexa Fluor« 350], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35154AF350
Product Code. 30499961
  • 6175.00 NOK / 0.10mL
Description
KLF1 Polyclonal antibody specifically detects KLF1 in Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KLF1
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Rat
Antibody
IgG
ELISA, Western Blot
Alexa Fluor 350
EKLFerythroid-specific transcription factor EKLF, Erythroid krueppel-like transcription factor, erythroid Kruppel-like factor, HBFQTL6, INLU, Krueppel-like factor 1, Kruppel-like factor 1 (erythroid), monoclonal A3D8
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KLF1 (NP_006554.1).,, Sequence:, MATAETALPSISTLTALGPFPDTQDDFLKWWRSEEAQDMGPGPPDPTEPPLHVKSEDQPGEEEDDERGADATWDLDLLLTNFSGPEPGGAPQTCALAPSE
0.1 mL
Cancer
10661
Store at 4°C in the dark.