Learn More
CACYBP Antibody (7D11A8) [Janelia Fluor« 669], Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-27912JF669
Product Code. 30499902
  • 7145.00 NOK / 0.10mL
Description
CACYBP Monoclonal antibody specifically detects CACYBP in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Serum
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CACYBP
Monoclonal
Janelia Fluor 669
CacyBP, calcyclin binding protein, GIG5, hCacyBP, MGC87971, RP1-102G20.6, S100A6-binding protein, S100A6BPgrowth-inhibiting gene 5 protein, Siah-interacting protein, Siah-interacting protein (SIP), SIPcalcyclin-binding protein
Truncated recombinant CACYBP expressed in E. Coli strain BL21 (DE3).
0.1 mL
Wnt Signaling Pathway
27101
Store at 4°C in the dark.
Purified
ELISA, Western Blot, Immunohistochemistry (Paraffin)
7D11A8
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Antibody
IgG1