missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR61 Antibody [DyLight 405], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-27894V
This item is not returnable.
View return policy
Description
WDR61 Polyclonal antibody specifically detects WDR61 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| WDR61 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 80349 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| DyLight 405 | |
| Meiotic recombination REC14 protein homolog, REC14, recombination protein REC14, SKI8, WD repeat domain 61, WD repeat-containing protein 61 | |
| WDR61 antibody was raised against a 14 amino acid peptide near the amino terminus of human WDR61. The immunogen is located within the first 50 amino acids of WDR61. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction