Learn More
RPS19 Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35925AF405
Product Code. 30499876
  • 6175.00 NOK / 0.10mL
Description
RPS19 Polyclonal antibody specifically detects RPS19 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPS19
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Antibody
IgG
ELISA, Western Blot
Alexa Fluor 405
DBADBA1,40S ribosomal protein S19, ribosomal protein S19
Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human RPS19 (NP_001013.1).,, Sequence:, MPGVTVKDVNQQEFVRALAAFLKKSGKLKVPEWVDTVKLAKHKELAPYDENWFYTRAASTARHLYLRGGAGVGSMTKIYGGRQRNGVMPSHFSRGSKSVARRVLQALEGLKMVEKDQDGGRKLTPQGQRDLDRIAGQVAAANKKH
0.1 mL
Core ESC Like Genes, Stem Cell Markers
6223
Store at 4°C in the dark.