missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin alpha 1 Antibody [DyLight 680], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-36712FR
This item is not returnable.
View return policy
Description
Spectrin alpha 1 Polyclonal antibody specifically detects Spectrin alpha 1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Spectrin alpha 1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| DyLight 680 | |
| alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 940-1160 of human Spectrin alpha 1 (NP_003117.2).,, Sequence:, KHEAFLLDLNSFGDSMKALRNQANACQQQQAAPVEGVAGEQRVMALYDFQARSPREVTMKKGDVLTLLSSINKDWWKVEAADHQGIVPAVYVRRLAHDEFPMLPQRRREEPGNITQRQEQIENQYRSLLDRAEERRRRLLQRYNEFLLAYEAGDMLEWIQEKKAENTGVELDDVWELQKKFDEFQKDLNTNEPRLRDINKVADDLLFEGLLTPEGAQIRQE | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 6708 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction