Learn More
SFXN1 Antibody [DyLight 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35295G
Product Code. 30499815
  • 6175.00 NOK / 0.10mL
Description
SFXN1 Polyclonal antibody specifically detects SFXN1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SFXN1
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
94081
Store at 4°C in the dark.
Purified
ELISA, Western Blot
DyLight 488
FLJ12876, sideroflexin 1, sideroflexin-1, TCC, Tricarboxylate carrier protein
Recombinant fusion protein containing a sequence corresponding to amino acids 87-322 of human SFXN1 (NP_073591.2).,, Sequence:, MILIGRMSAQVPMNMTITGCMMTFYRTTPAVLFWQWINQSFNAVVNYTNRSGDAPLTVNELGTAYVSATTGAVATALGLNALTKHVSPLIGRFVPFAAVAAANCINIPLMRQRELKVGIPVTDENGNRLGESANAAKQAITQVVVSRILMAAPGMAIPPFIMNTLEKKAFLKRFPWMSAPIQVGLVGFCLVFATPLCCALFPQKSSMSVTSLEAELQAKIQESHPELRRVYFNKGL
0.1 mL
Primary
Human, Mouse, Rat