missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthine Oxidase Antibody [FITC], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35302F
This item is not returnable.
View return policy
Description
Xanthine Oxidase Polyclonal antibody specifically detects Xanthine Oxidase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Xanthine Oxidase | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| FITC | |
| EC 1.17.1.4, EC 1.7.2.2, xanthene dehydrogenase, xanthine dehydrogenase, xanthine dehydrogenase/oxidase, xanthine oxidase, xanthine oxidoreductase, XDHA, XO, XOR | |
| A synthetic peptide corresponding to a sequence within amino acids 985-1084 of human Xanthine Oxidase (XDH) (NP_000370.2).,, Sequence:, CIIPTKFGISFTVPFLNQAGALLHVYTDGSVLLTHGGTEMGQGLHTKMVQVASRALKIPTSKIYISETSTNTVPNTSPTAASVSADLNGQAVYAACQTILK | |
| 0.1 mL | |
| Signal Transduction | |
| 7498 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction