missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 5/CHRNA5 Antibody [Alexa Fluor« 350], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35746AF350
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 5/CHRNA5 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 5/CHRNA5 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Nicotinic Acetylcholine R alpha 5/CHRNA5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Alexa Fluor 350 | |
| Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5, cholinergic receptor, nicotinic, alpha 5, cholinergic receptor, nicotinic, alpha polypeptide 5, LNCR2, NACHRA5, neuronal acetylcholine receptor subunit alpha-5, neuronal nicotinic acetylcholine receptor, alpha5 subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-240 of human Nicotinic Acetylcholine R alpha 5/CHRNA5 (NP_000736.2).,, Sequence:, RCGLAGAAGGAQRGLSEPSSIAKHEDSLLKDLFQDYERWVRPVEHLNDKIKIKFGLAISQLVDVDEKNQLMTTNVWLKQEWIDVKLRWNPDDYGGIKVIRVPSDSVWTPDIVLFDNADGRFEGTSTKTVIRYNGTVTWTPPANYKSSCTIDVTFFPFDLQNCSMKFGSWTYDGSQVDIILEDQDVDKRDFFDNGEWEIVSATGSKGNRTDSCCWYPYV | |
| 0.1 mL | |
| Signal Transduction | |
| 1138 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction