missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUCY2D Antibody [HRP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35345H
This item is not returnable.
View return policy
Description
GUCY2D Polyclonal antibody specifically detects GUCY2D in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| GUCY2D | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| HRP | |
| CORD6cone rod dystrophy 5/6, CYGD, EC 4.6.1, guanylate cyclase 2D, membrane (retina-specific), Guanylate cyclase 2D, retinal, GUC1A4, GUC2DCORD5, LCA, LCA1RCD2, retGC, RETGC1, RETGC-1EC 4.6.1.2, retinal guanylyl cyclase 1, Rod outer segment membrane guanylate cyclase, ROSGC, ROS-GC, ROS-GC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 52-350 of human GUCY2D (NP_000171.1).,, Sequence:, AVFTVGVLGPWACDPIFSRARPDLAARLAAARLNRDPGLAGGPRFEVALLPEPCRTPGSLGAVSSALARVSGLVGPVNPAACRPAELLAEEAGIALVPWGCPWTQAEGTTAPAVTPAADALYALLRAFGWARVALVTAPQDLWVEAGRSLSTALRARGLPVASVTSMEPLDLSGAREALRKVRDGPRVTAVIMVMHSVLLGGEEQRYLLEAAEELGLTDGSLVFLPFDTIHYALSPGPEALAALANSSQLRRAHDAVLTLTRHCPSEGSVLDSLRRAQERRELPSDLNLQQVSPLFGTI | |
| 0.1 mL | |
| Protein Kinase | |
| 3000 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction