Learn More
GADD45G Antibody [Janelia Fluor« 525], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35120JF525
Product Code. 30499769
  • 6175.00 NOK / 0.10mL
Description
GADD45G Polyclonal antibody specifically detects GADD45G in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GADD45G
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Mouse, Rat
Antibody
IgG
ELISA, Western Blot
Janelia Fluor 525
CR6DNA damage-inducible transcript 2 protein, Cytokine-responsive protein CR6, DDIT-2, DDIT2GADD45-gamma, growth arrest and DNA damage-inducible protein GADD45 gamma, growth arrest and DNA-damage-inducible, gamma, GRP1717 kD
Recombinant fusion protein containing a sequence corresponding to amino acids 90-159 of human GADD45G (NP_006696.1).,, Sequence:, IDIVRVGDVQRLAAIVGAGEEAGAPGDLHCILISNPNEDAWKDPALEKLSLFCEESRSVNDWVPSITLPE
0.1 mL
Apoptosis, Cancer
10912
Store at 4°C in the dark.