missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 20S beta 6 Antibody [PerCP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38072PCP
This item is not returnable.
View return policy
Description
Proteasome 20S beta 6 Polyclonal antibody specifically detects Proteasome 20S beta 6 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Proteasome 20S beta 6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| PerCP | |
| DELTA, EC 3.4.25.1, LMPY, Macropain delta chain, Multicatalytic endopeptidase complex delta chain, proteasome (prosome, macropain) subunit, beta type, 6, proteasome catalytic subunit 1, Proteasome delta chain, proteasome subunit beta 6, proteasome subunit beta type-6, proteasome subunit delta, Proteasome subunit Y, PSY large multifunctional protease Y, YMGC5169 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-239 of human Proteasome 20S beta 6 (NP_002789.1).,, Sequence:, TTIMAVQFDGGVVLGADSRTTTGSYIANRVTDKLTPIHDRIFCCRSGSAADTQAVADAVTYQLGFHSIELNEPPLVHTAASLFKEMCYRYREDLMAGIIIAGWDPQEGGQVYSVPMGGMMVRQSFAIGGSGSSYIYGYVDATYREGMTKEECLQFTANALALAMERDGSSGGVIRLAAIAESGVERQVLLGDQIPKFAVATLPPA | |
| 0.1 mL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
| 5694 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction