missing translation for 'onlineSavingsMsg'
Learn More

Karyopherin (importin) beta 3 Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™

Product Code. 30499059 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30499059 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30499059

Brand: Novus Biologicals NBP335908AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Karyopherin (importin) beta 3 Polyclonal antibody specifically detects Karyopherin (importin) beta 3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Karyopherin (importin) beta 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias DKFZp686O1576, FLJ43041, IMB3, imp5, importin 5, importin beta-3 subunit, Importin subunit beta-3, importin-5, karyopherin (importin) beta 3, Karyopherin beta-3, KPNB3, MGC2068, RAN binding protein 5, Ran_GTP binding protein 5, Ran-binding protein 5, RANBP5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human Karyopherin (importin) beta 3 (NP_002262.3).,, Sequence:, AAAAAEQQQFYLLLGNLLSPDNVVRKQAEETYENIPGQSKITFLLQAIRNTTAAEEARQMAAVLLRRLLSSAFDEVYPALPSDVQTAIKSELLMIIQMETQSSMRKKVCDIAAELARNLIDEDGNNQWPEGLKFLFDSVSSQNVGLREAALHIFWNFPGIFGNQQQHYLDVIKRMLVQCMQDQEHPSIRTLSARATAAFILANEHNVALFKHFADLLPGFLQAVNDSCYQNDDSVLKSLVEIADTVPKYLRPHLEATLQLSLKLCGDTSLNNMQRQLALEV
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3843
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.