missing translation for 'onlineSavingsMsg'
Learn More

DNAJC7 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30499036 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30499036 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30499036

Brand: Novus Biologicals NBP338013AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNAJC7 Polyclonal antibody specifically detects DNAJC7 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias DANJC7, DJ11, DnaJ (Hsp40) homolog, subfamily C, member 7, tetratricopeptide repeat domain 2, Tetratricopeptide repeat protein 2, TPR repeat protein 2, TPR2DJC7, TTC2dnaJ homolog subfamily C member 7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 230-380 of human DNAJC7 (NP_003306.3).,, Sequence:, VQFFVQALRMAPDHEKACIACRNAKALKAKKEDGNKAFKEGNYKLAYELYTEALGIDPNNIKTNAKLYCNRGTVNSKLRKLDDAIEDCTNAVKLDDTYIKAYLRRAQCYMDTEQYEEAVRDYEKVYQTEKTKEHKQLLKNAQLELKKSKRK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7266
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.