missing translation for 'onlineSavingsMsg'
Learn More

NOP58 Antibody [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30499007 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30499007 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30499007

Brand: Novus Biologicals NBP338111APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOP58 Polyclonal antibody specifically detects NOP58 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP58
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias HSPC120, NOL5, NOP5/NOP58, NOP58 ribonucleoprotein homolog (yeast), NOP5nucleolar protein 58, Nucleolar protein 5, nucleolar protein NOP5/NOP58
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 360-529 of human NOP58 (NP_057018.1).,, Sequence:, RMLAAKTVLAIRYDAFGEDSSSAMGVENRAKLEARLRTLEDRGIRKISGTGKALAKTEKYEHKSEVKTYDPSGDSTLPTCSKKRKIEQVDKEDEITEKKAKKAKIKVKVEEEEEEKVAEEEETSVKKKKKRGKKKHIKEEPLSEEEPCTSTAIASPEKKKKKKKKRENED
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 51602
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.